Silver sulphate, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ100
497266-10G 497266-50G
SIAL497266-10GEA 91.5 EUR
SIAL497266-10G SIAL497266-50G
Silver sulphate, Sigma-Aldrich®
Silver sulphate
Formula: Ag₂SO₄
Boiling Pt: 1085 °C (1013 hPa)
Melting Pt: 655 °C
Density: 5.45 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003407
CAS Number: 10294-26-5
EINECS: 233-653-7
Merck Index: 13,08603
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dd7afaa8e9ae24bf040d40aed0c93763ce897080&vid=ae7b50634b53507047974c2157e5e38c2eb90a20&prid=web&clid=SAPDAM