Zinc bromide, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Zinc (II) bromide
M-Clarity™ quality level = MQ100
230022-10G 230022-50G
SIAL230022-10GEA 85.2 EUR
SIAL230022-10G SIAL230022-50G
Zinc bromide, Sigma-Aldrich®
Zinc bromide
Formula: Br₂Zn
Boiling Pt: 670…680 °C (1013 hPa)
Melting Pt: 394 °C
Density: 4.22 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011294
CAS Number: 7699-45-8
EINECS: 231-718-4
UN: 3260
ADR: 8,II
Merck Index: 13,10182