Zinc bromide, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
Zinc (II) bromide
M-Clarity™ quality level = MQ100
451398-25G
451398-5G
SIAL451398-25GEA
359
EUR
SIAL451398-25G
SIAL451398-5G
Zinc bromide, Sigma-Aldrich®
Zinc bromide
Formula:
Br₂Zn Boiling Pt: 670…680 °C (1013 hPa) Melting Pt: 394 °C Density: 4.22 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011294 CAS Number: 7699-45-8 EINECS: 231-718-4 UN: 3260 ADR: 8,II Merck Index: 13,10182 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...