Cadmium(II) chloride, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Cadmium chloride
M-Clarity™ quality level = MQ100
655198-100G 655198-5G
SIAL655198-100GEA 160 EUR
SIAL655198-100G SIAL655198-5G
Cadmium(II) chloride, Sigma-Aldrich®
Cadmium (II) chloride
Formula: CdCl₂
MW: 183.32 g/mol
Boiling Pt: 967
Melting Pt: 565
Density: 4.05 g/cm³ (20 °C)
MDL Number: MFCD00010916
CAS Number: 10108-64-2
EINECS: 233-296-7
UN: 2570
ADR: 6.1 III
Merck Index: 12,01653