Zinc chloride, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Zinc dichloride, Dichlorozinc, Zinc (II) chloride
M-Clarity™ quality level = MQ100
450111-10G 450111-50G
SIAL450111-10GEA 246 EUR
SIAL450111-10G SIAL450111-50G
Zinc chloride, Sigma-Aldrich®
Zinc chloride
Formula: ZnCl₂
Boiling Pt: 732 °C (1013 hPa)
Melting Pt: 283 °C
Density: 2.91 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011295
CAS Number: 7646-85-7
EINECS: 231-592-0
UN: 2331
ADR: 8,III
Merck Index: 13,10185