Zinc chloride, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: Zinc dichloride, Dichlorozinc, Zinc (II) chloride

M-Clarity™ quality level = MQ200

456845-25G 456845-5G
SIAL456845-25GEA 443 EUR
SIAL456845-25G SIAL456845-5G
Zinc chloride, Sigma-Aldrich®
Zinc chloride
Formula: ZnCl₂
MW: 136.3 g/mol
Boiling Pt: 732
Melting Pt: 283
Density: 2.91 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011295
CAS Number: 7646-85-7
EINECS: 231-592-0
UN: 2331
ADR: 8 III
Merck Index: 13,10185

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR