Zinc chloride, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
M-Clarity™ quality level = MQ200
Z0152-100G
Z0152-1KG
Z0152-500G
SIALZ0152-100GEA
33.7
EUR
SIALZ0152-100G
SIALZ0152-1KG
SIALZ0152-500G
Zinc chloride, Sigma-Aldrich®
Zinc chloride
Formula:
ZnCl₂ Boiling Pt: 732 °C (1013 hPa) Melting Pt: 283 °C Density: 2.91 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011295 CAS Number: 7646-85-7 EINECS: 231-592-0 UN: 2331 ADR: 8,III Merck Index: 13,10185 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...