Zinc iodide 98+%, pure
Supplier: Thermo Fisher Scientific
Danger
Synonyms: Zinc (II) iodide
208060500 208062500 208060010
ACRO208060500EA 59.3 EUR
ACRO208060500 ACRO208062500 ACRO208060010
Zinc iodide 98+%, pure
Zinc iodide
Formula: I₂Zn
Boiling Pt: 620 °C (1013 hPa)
Melting Pt: 446 °C
Density: 4.74 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011299
CAS Number: 10139-47-6
EINECS: 233-396-0
UN: 3260
ADR: 8,II
Merck Index: 12,10269
Order Now
Specification Test Results
Appearance White to yellow to light beige Hygroscopic crystalline powder or crystals
Titration Argentometric ≥98.0 %