Zinc iodide, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ200
223883-250G 223883-50G
SIAL223883-250GEA 222 EUR
SIAL223883-250G SIAL223883-50G
Zinc iodide, Sigma-Aldrich®
Zinc iodide
Formula: I₂Zn
Boiling Pt: 620 °C (1013 hPa)
Melting Pt: 446 °C
Density: 4.74 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011299
CAS Number: 10139-47-6
EINECS: 233-396-0
UN: 3260
ADR: 8,II
Merck Index: 12,10269