Cadmium oxide 99%, pure
Supplier: Thermo Fisher Scientific
Danger
223790010 223790050 223792500
ACRO223790010EA 108 EUR
ACRO223790010 ACRO223790050 ACRO223792500
Cadmium oxide 99%, pure
Cadmium oxide
Formula: CdO
Boiling Pt: 1385 °C (760 mmHg)
Melting Pt: ~ 1426 °C
Density: 8,15 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010921
CAS Number: 1306-19-0
EINECS: 215-146-2
UN: 2570
ADR: 6.1,I
Merck Index: 13,01623
Order Now
Specification Test Results
Appearance Red-brown to brown Powder
Titration Complexometric ≥98.5 %