Zinc selenide, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Zinc (II) selenide
M-Clarity™ quality level = MQ100
244619-10G 244619-50G
SIAL244619-10GEA 101 EUR
SIAL244619-10G SIAL244619-50G
Zinc selenide, Sigma-Aldrich®
Zinc selenide
Formula: SeZn
MW: 144.35 g/mol
Melting Pt: 1793
Density: 5.4 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011296
CAS Number: 1315-09-9
EINECS: 215-259-7
UN: 3283
ADR: 6.1 III
Merck Index: 12,10290