Discandium trioxide ≥99.99% (REO, rare earth oxide basis), REacton®
Supplier: Thermo Fisher Scientific
Synonyms: Scandium oxide, Scandium (III) oxide
11217.03 11217.06 11217.14
11217.03EA 123 EUR
11217.03 11217.06 11217.14
Discandium trioxide ≥99.99% (REO, rare earth oxide basis), REacton®
Discandium trioxide
Purity: 99.99% CAS No.: 12060-08-1 Molecular Formula: Sc2O3 Formula Weight: 137.91 Physical Form: Powder Refractive Index: 1.964 Odour: Odourless Melting Point: 2485° Merck Reference: 14,8392 MDL No.: MFCD00011223
Formula: O₃Sc₂
MW: 137.91 g/mol
Melting Pt: 2485 °C
Density: 3.86 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011223
CAS Number: 12060-08-1
EINECS: 235-042-0
Merck Index: 14,8392