Zirconium tetrachloride, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: Zirconium (IV) chloride

M-Clarity™ quality level = MQ100

647640-25G 647640-5G
SIAL647640-25GEA 442 EUR
SIAL647640-25G SIAL647640-5G
Zirconium tetrachloride, Sigma-Aldrich®
Zirconium tetrachloride
Formula: Cl₄Zr
Boiling Pt: 3577 °C (1013 hPa)
Melting Pt: 437 °C
Density: 2.8 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011306
CAS Number: 10026-11-6
EINECS: 233-058-2
UN: 2503
ADR: 8,III
Merck Index: 12,10307

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR