Titanium(III) oxide ≥99.8% (metals basis)
Supplier: Thermo Fisher Scientific
77127.18 77127.30
77127.18EA 200 EUR
77127.18 77127.30
Titanium(III) oxide ≥99.8% (metals basis)
Titanium (III) oxide
Purity: 99.8% CAS No.: 1344-54-3 Molecular Formula: Ti2O3 Formula Weight: 143.80 Physical Form: -325 Mesh Powder Density: 4.6 Melting Point: 2130° dec. MDL No.: MFCD00036278
Formula: O₃Ti₂
MW: 143.76 g/mol
Melting Pt: 2130 °C (dec.)
Density: 4.6
Storage Temperature: Ambient
MDL Number: MFCD00036278
CAS Number: 1344-54-3
EINECS: 215-697-9