Zirconium(IV) oxide, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Zirconium dioxide, Zirconium Oxide
M-Clarity™ quality level = MQ200
230693-100G 230693-2KG 230693-500G
SIAL230693-100GEA 59.5 EUR
SIAL230693-100G SIAL230693-2KG SIAL230693-500G
Zirconium(IV) oxide, Sigma-Aldrich®
Zirconium (IV) oxide
Formula: ZrO₂
MW: 123.22 g/mol
Boiling Pt: 4300
Melting Pt: 2680
Density: 5.75 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011310
CAS Number: 1314-23-4
EINECS: 215-227-2
Merck Index: 14,10180