Titanium(IV) isopropoxide, Sigma-Aldrich®
Supplier: Merck
Synonyms: Titanium tetraisopropanolate, Titanium (IV) isopropoxide
MQ200
377996-100ML 377996-25ML 377996-5ML
SIAL377996-100MLEA 467 EUR
SIAL377996-100ML SIAL377996-25ML SIAL377996-5ML
Titanium(IV) isopropoxide, Sigma-Aldrich®
Titanium (IV) isopropoxide
Formula: C₁₂H₂₈O₄Ti
MW: 284.23 g/mol
Boiling Pt: 240
Melting Pt: 18…20 °C
Density: 0.9711 g/cm³ (20 °C)
Flash Pt: 51
MDL Number: MFCD00008871
CAS Number: 546-68-9
EINECS: 208-909-6
UN: 2413
ADR: 3 III
Merck Index: 12,09614