Titanium(IV) propanolate, Sigma-Aldrich®
Supplier: Merck
Synonyms: Tetrapropyl orthotitanate, Titanium (IV) propoxide, Titanium tetrapropanolate
M-Clarity™ quality level = MQ200
253081-100G 253081-500G
SIAL253081-100GEA 63.5 EUR
SIAL253081-100G SIAL253081-500G
Titanium(IV) propanolate, Sigma-Aldrich®
Titanium(IV) propanolate
Formula: C₁₂H₂₈O₄Ti
Boiling Pt: 137 °C (7 hPa)
Density: 1.033 g/cm³ (20 °C)
MDL Number: MFCD00009365
CAS Number: 3087-37-4
EINECS: 221-411-3
UN: 2413
ADR: 3,III