Niobium(V) oxide, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
203920-10G 203920-250G 203920-50G
SIAL203920-10GEA 104 EUR
SIAL203920-10G SIAL203920-250G SIAL203920-50G
Niobium(V) oxide, Sigma-Aldrich®
Niobium (V) oxide
Formula: Nb₂O₅
Melting Pt: 1496 °C
Density: 4.6 g/cm³ (20 °C)
MDL Number: MFCD00011128
CAS Number: 1313-96-8
EINECS: 215-213-6
Merck Index: 14,06559