Tantalum(V) oxide, Sigma-Aldrich®
Supplier: Merck
Synonyms: Tantalum(V) oxide, Ditantalum pentaoxide
M-Clarity™ quality level = MQ100
204536-50G 204536-10G
SIAL204536-50GEA 357 EUR
SIAL204536-50G SIAL204536-10G
Tantalum(V) oxide, Sigma-Aldrich®
Tantalum (V) oxide
Formula: O₅Ta₂
MW: 441.89 g/mol
Melting Pt: 1870…1880 °C
Density: 8.2 g/cm³ (20 °C)
MDL Number: MFCD00011254
CAS Number: 1314-61-0
EINECS: 215-238-2
Merck Index: 14,09056