Tantalum(V) oxide, Sigma-Aldrich®

Supplier: Merck

Synonyms: Tantalum(V) oxide, Ditantalum pentaoxide

M-Clarity™ quality level = MQ100

303518-100G 303518-25G
SIAL303518-100GEA 451 EUR
SIAL303518-100G SIAL303518-25G
Tantalum(V) oxide, Sigma-Aldrich®
Tantalum (V) oxide
Formula: O₅Ta₂
Melting Pt: 1870…1880 °C
Density: 8.2 g/cm³ (20 °C)
MDL Number: MFCD00011254
CAS Number: 1314-61-0
EINECS: 215-238-2
Merck Index: 14,09056

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR