Chromium(II) chloride, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Chromium dichloride
M-Clarity™ quality level = MQ200
244805-25G 244805-5G
SIAL244805-25GEA 586 EUR
SIAL244805-25G SIAL244805-5G
Chromium(II) chloride, Sigma-Aldrich®
Chromium (II) chloride
Formula: Cl₂Cr
MW: 122.9 g/mol
Boiling Pt: 1302
Melting Pt: 823…825 °C
Density: 2.87 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010947
CAS Number: 10049-05-5
EINECS: 233-163-3
UN: 3260
ADR: 8 III
Merck Index: 13,02264