Tungsten(VI) chloride, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: Tungsten hexachloride

M-Clarity™ quality level = MQ100

241911-100G 241911-10G
SIAL241911-100GEA 473 EUR
SIAL241911-100G SIAL241911-10G
Tungsten(VI) chloride, Sigma-Aldrich®
Tungsten (VI) chloride
Formula: Cl₆W
MW: 396.56 g/mol
Boiling Pt: 347
Melting Pt: 275
Density: 3.518 g/cm³ (20 °C)
MDL Number: MFCD00011463
CAS Number: 13283-01-7
EINECS: 236-293-9
UN: 3260
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR