Chromium(VI) oxide, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
Chromic anhydride, Chromium trioxide
M-Clarity™ quality level = MQ200
675644-25G
675644-5G
SIAL675644-25GEA
108
EUR
SIAL675644-25G
SIAL675644-5G
Chromium(VI) oxide, Sigma-Aldrich®
Chromium (VI) oxide
Formula:
CrO₃ MW: 99.99 g/mol Melting Pt: 195 Density: 2.7 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010952 CAS Number: 1333-82-0 EINECS: 215-607-8 UN: 1463 ADR: 5.1 II REACH: 01-2119458868-17 Merck Index: 13,02256 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...