Tungsten trioxide, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: Tungsten (VI) oxide

M-Clarity™ quality level = MQ200

95410-100G 95410-500G
SIAL95410-100GEA 108 EUR
SIAL95410-100G SIAL95410-500G
Tungsten trioxide, Sigma-Aldrich®
Tungsten trioxide
Formula: O₃W
MW: 231.84 g/mol
Melting Pt: 1473
Density: 7.16 g/cm³ (20 °C)
MDL Number: MFCD00011466
CAS Number: 1314-35-8
EINECS: 215-231-4
Merck Index: 12,09947

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR