Rhenium trichloride, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
M-Clarity™ quality level = MQ100
309184-1G
309184-5G
SIAL309184-1GEA
478
EUR
SIAL309184-1G
SIAL309184-5G
Rhenium trichloride, Sigma-Aldrich®
Rhenium trichloride
Formula:
Cl₃Re |
MDL Number:
MFCD00011196 CAS Number: 13569-63-6 EINECS: 236-987-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...