Rhenium pentachloride, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: Rhenium(V) chloride

M-Clarity™ quality level = MQ100

309192-1G 309192-5G
SIAL309192-1GEA 255 EUR
SIAL309192-1G SIAL309192-5G
Rhenium pentachloride, Sigma-Aldrich®
Rhenium pentachloride
Formula: Cl₅Re
MW: 363.47 g/mol
MDL Number: MFCD00011197
CAS Number: 13596-35-5
EINECS: 237-042-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR