Iron(II) bromide, Sigma-Aldrich®

Supplier: Merck

Synonyms: Iron dibromide, Ferrous bromide, Iron(II) bromide

M-Clarity™ quality level = MQ100

400831-10G 400831-50G
SIAL400831-10GEA 87.9 EUR
SIAL400831-10G SIAL400831-50G
Iron(II) bromide, Sigma-Aldrich®
Iron (II) bromide
Formula: Br₂Fe
MW: 215.66 g/mol
MDL Number: MFCD00016081
CAS Number: 7789-46-0
EINECS: 232-168-8
UN: 3260
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR