Nickel(II) bromide, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: Nickel dibromide

M-Clarity™ quality level = MQ100

561142-10G 561142-2G
SIAL561142-10GEA 280 EUR
SIAL561142-10G SIAL561142-2G
Nickel(II) bromide, Sigma-Aldrich®
Nickel (II) bromide
Formula: Br₂Ni
MW: 218.5 g/mol
Melting Pt: 963
Density: 5.09 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011141
CAS Number: 13462-88-9
EINECS: 236-665-0
UN: 3077
ADR: 9 III
Merck Index: 12,06585

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR