Cobalt(II) chloride, anhydrous 97%
Supplier: Thermo Fisher Scientific
Danger
Synonyms: Cobaltous chloride, Cobalt dichloride
B22031.14 B22031.22 B22031.36
B22031.14EA 37.6 EUR
B22031.14 B22031.22 B22031.36
Cobalt(II) chloride, anhydrous 97%
Cobalt (II) chloride
Purity: 97% CAS No.: 7646-79-9 Molecular Formula: Cl2Co Formula Weight: 129.84 Odour: Odourless Melting Point: 735° Boiling Point: 1049° Merck Reference: 14,2437 UN No.: 3077 MDL No.: MFCD00010938
Formula: Cl₂Co
MW: 129.84 g/mol
Boiling Pt: 1049 °C
Melting Pt: 735
Density: 3.367
Storage Temperature: Ambient
MDL Number: MFCD00010938
CAS Number: 7646-79-9
EINECS: 231-589-4
UN: 3077
ADR: 9 III
Merck Index: 14,2437