Cobalt(II) chloride, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Cobaltous chloride, Cobalt dichloride
M-Clarity™ quality level = MQ200
409332-1G 409332-5G
SIAL409332-1GEA 114 EUR
SIAL409332-1G SIAL409332-5G
Cobalt(II) chloride, Sigma-Aldrich®
Cobalt (II) chloride
Formula: Cl₂Co
MW: 129.84 g/mol
Boiling Pt: 1049
Melting Pt: 735
Density: 1.6795 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00010938
CAS Number: 7646-79-9
EINECS: 231-589-4
UN: 3077
ADR: 9 III
Merck Index: 12,02498