Iron(II) chloride, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Iron dichloride, Ferrous chloride
M-Clarity™ quality level = MQ100
372870-250G 372870-25G
SIAL372870-250GEA 707 EUR
SIAL372870-250G SIAL372870-25G
Iron(II) chloride, Sigma-Aldrich®
Iron (II) chloride
Formula: Cl₂Fe
MW: 126.75 g/mol
Boiling Pt: 2023
Melting Pt: 674
Density: 2.7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011004
CAS Number: 7758-94-3
EINECS: 231-843-4
UN: 3260
ADR: 8 III
Merck Index: 12,04091