Iron(III) chloride, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Iron trichloride, Ferric chloride
M-Clarity™ quality level = MQ100
451649-1G 451649-5G
SIAL451649-1GEA 179 EUR
SIAL451649-1G SIAL451649-5G
Iron(III) chloride, Sigma-Aldrich®
Iron (III) chloride
Formula: FeCl₃
MW: 162.21 g/mol
Boiling Pt: 315
Melting Pt: 260
Density: 2.8 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011005
CAS Number: 7705-08-0
EINECS: 231-729-4
UN: 1773
ADR: 8 III
Merck Index: 13,04048