Iron(III) chloride, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Iron trichloride, Ferric chloride
M-Clarity™ quality level = MQ100
701122-1G 701122-25G 701122-5G
SIAL701122-1GEA 37.1 EUR
SIAL701122-1G SIAL701122-25G SIAL701122-5G
Iron(III) chloride, Sigma-Aldrich®
Iron (III) chloride
Formula: FeCl₃
MW: 162.21 g/mol
Boiling Pt: 315
Melting Pt: 260
Density: 2.8 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011005
CAS Number: 7705-08-0
EINECS: 231-729-4
UN: 1773
ADR: 8 III
Merck Index: 13,04048