Nickel(II) chloride, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Nickel dichloride
M-Clarity™ quality level = MQ100
339350-250G 339350-50G
SIAL339350-250GEA 294 EUR
SIAL339350-250G SIAL339350-50G
Nickel(II) chloride, Sigma-Aldrich®
Nickel (II) chloride
Formula: Cl₂Ni
MW: 129.6 g/mol
Melting Pt: 1009
Density: 3.55 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011142
CAS Number: 7718-54-9
EINECS: 231-743-0
UN: 3288
ADR: 6.1 III
Merck Index: 12,06588