Nickel(II) chloride, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Nickel dichloride
M-Clarity™ quality level = MQ200
451193-25G 451193-5G
SIAL451193-25GEA 779 EUR
SIAL451193-25G SIAL451193-5G
Nickel(II) chloride, Sigma-Aldrich®
Nickel (II) chloride
Formula: Cl₂Ni
MW: 129.6 g/mol
Melting Pt: 1009
Density: 3.55 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011142
CAS Number: 7718-54-9
EINECS: 231-743-0
UN: 3288
ADR: 6.1 III
Merck Index: 12,06588