Platinum(IV) chloride, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Platinum tetrachloride
M-Clarity™ quality level = MQ100
520640-1G 520640-5G
SIAL520640-1GEA 221 EUR
SIAL520640-1G SIAL520640-5G
Platinum(IV) chloride, Sigma-Aldrich®
Platinum (IV) chloride
Formula: Cl₄Pt
MW: 336.89 g/mol
Melting Pt: 370
Density: 4.3 g/cm³ (20 °C)
MDL Number: MFCD00011182
CAS Number: 13454-96-1
EINECS: 236-645-1
UN: 3260
ADR: 8 III