Cobalt(III) fluoride, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
Cobaltic fluoride, Cobalt trifluoride
M-Clarity™ quality level = MQ100
236136-25G
236136-5G
SIAL236136-25GEA
107
EUR
SIAL236136-25G
SIAL236136-5G
Cobalt(III) fluoride, Sigma-Aldrich®
Cobalt (III) fluoride
Formula:
CoF₃ Melting Pt: 927 °C Density: 3.88 g/cm³ (20 °C) |
MDL Number:
MFCD00010942 CAS Number: 10026-18-3 EINECS: 233-062-4 UN: 3260 ADR: 8,II Merck Index: 12,02491 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...