Cobalt(II,III) oxide, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
Synonyms:
Tricobalt tetraoxide, Cobaltic tetraoxide
M-Clarity™ quality level = MQ200
637025-100G
637025-250G
637025-25G
SIAL637025-100GEA
308
EUR
SIAL637025-100G
SIAL637025-250G
SIAL637025-25G
Cobalt(II,III) oxide, Sigma-Aldrich®
Cobalt (II,III) oxide
Formula:
Co₃O₄ MW: 240.8 g/mol Melting Pt: 895 Density: 6.005 g/cm³ (20 °C) |
MDL Number:
MFCD00010939 CAS Number: 1308-06-1 EINECS: 215-157-2 Merck Index: 13,02454 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...