Iron(II,III) oxide, Sigma-Aldrich®

Supplier: Merck
Warning

Synonyms: Magnetite, Triiron tetraoxide, Iron(II,III) oxide

M-Clarity™ quality level = MQ100

637106-100G 637106-250G 637106-25G
SIAL637106-100GEA 251 EUR
SIAL637106-100G SIAL637106-250G SIAL637106-25G
Iron(II,III) oxide, Sigma-Aldrich®
Iron (II,III) oxide
Formula: Fe₃O₄
MW: 231.54 g/mol
Melting Pt: 1560
Density: 5.18 g/cm³ (20 °C)
MDL Number: MFCD00011010
CAS Number: 1317-61-9
EINECS: 215-277-5
Merck Index: 12,04087

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR