Nickel (II) oxide, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Nickel monoxide, Nickel oxide, Bunsenite
M-Clarity™ quality level = MQ100
203882-100G 203882-20G
SIAL203882-100GEA 659 EUR
SIAL203882-100G SIAL203882-20G
Nickel (II) oxide, Sigma-Aldrich®
Nickel (II) oxide
Formula: NiO
Melting Pt: 1990 °C
Density: 4.6…6.67 g/cm³ (20 °C)
MDL Number: MFCD00011145
CAS Number: 1313-99-1
EINECS: 215-215-7