Cobalt(II) hydroxide ≥97%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: Cobalt dihydroxide, Cobaltous hydroxide, Cobalt(II) hydroxide
B22045.18 B22045.30
B22045.18EA 34.9 EUR
B22045.18 B22045.30
Cobalt(II) hydroxide ≥97%
Cobalt (II) hydroxide
Purity: 97% CAS No.: 21041-93-0 Molecular Formula: H2CoO2 Formula Weight: 92.95 Odour: Odourless Melting Point: 250° dec. MDL No.: MFCD00016015
Formula: H₂CoO₂
MW: 92.95 g/mol
Melting Pt: 250 °C (dec.)
Density: 3.597 g/cm³ (15 °C)
Storage Temperature: Ambient
MDL Number: MFCD00016015
CAS Number: 21041-93-0
EINECS: 244-166-4
Merck Index: 13,02467