γ-Butyrolactone, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: gamma-Butyrolactone, Butan-4-olide, 4-Hydroxybutanoic acid lactone, 1,4-Butyrolactone, GBL
M-Clarity™ quality level = MQ200
B103608-20KG B103608-25G B103608-500G B103608-2.5KG
SIALB103608-20KGEA 670 EUR
SIALB103608-20KG SIALB103608-25G SIALB103608-500G SIALB103608-2.5KG
γ-Butyrolactone, Sigma-Aldrich®
γ-Butyrolactone
Formula: C₄H₆O₂
Boiling Pt: 204 °C (1013 hPa)
Melting Pt: –44 °C
Density: 1.1286 g/cm³ (20 °C)
Flash Pt: 104 °C
Storage Temperature: Ambient
MDL Number: MFCD00005386
CAS Number: 96-48-0
EINECS: 202-509-5
Merck Index: 13,01596
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e855ee75d3243dd31c0ed64e2354b467a7de1f40&vid=a50dd849ab627dd2dde8674edf04ce9bad83f843&prid=web&clid=SAPDAM