Sorbitan monolaurate
Supplier: Thermo Fisher Scientific
Synonyms: Sorbitan monododecanoate, Sorbitan laurate, Span 20
L12099.30 L12099.0B
L12099.30EA 38.9 EUR
L12099.30 L12099.0B
Sorbitan monolaurate
Sorbitan monolaurate
CAS No.: 1338-39-2 Molecular Formula: C18H34O6 Formula Weight: 346.47 Flash Point: 110°(230°F) Refractive Index: 1.4740 Merck Reference: 14,8724 MDL No.: MFCD00005365
Formula: C₁₈H₃₄O₆
MW: 346.46 g/mol
Density: 1.032
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005365
CAS Number: 1338-39-2
EINECS: 215-663-3
Merck Index: 14,8724