9-Bromophenanthrene ≥98%

Supplier: Thermo Fisher Scientific
Danger

L09033.06 L09033.14
L09033.06EA 25.7 EUR
L09033.06 L09033.14
9-Bromophenanthrene ≥98%
9-Bromophenanthrene

Purity: 98% CAS No.: 573-17-1 Molecular Formula: C14H9Br Formula Weight: 257.14 Melting Point: 62-66° Boiling Point: 190°/1.2mm MDL No.: MFCD00001174

Formula: C₁₄H₉Br
MW: 257.13 g/mol
Boiling Pt: 190 °C (1.2 mmHg)
Melting Pt: 62…66 °C
Density: 1.4093 g/cm³ (101 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001174
CAS Number: 573-17-1
EINECS: 209-351-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR