9-Bromophenanthrene, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
B75409-10G
B75409-25G
SIALB75409-10GEA
63
EUR
SIALB75409-10G
SIALB75409-25G
9-Bromophenanthrene, Sigma-Aldrich®
9-Bromophenanthrene
Formula:
C₁₄H₉Br Boiling Pt: 390 °C (1013 hPa) Melting Pt: 62 °C Density: 1.4093 g/cm³ (101 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00001174 CAS Number: 573-17-1 EINECS: 209-351-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...