Phenanthrene, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
M-Clarity™ quality level = MQ100
695114-1G
695114-5G
SIAL695114-1GEA
52
EUR
SIAL695114-1G
SIAL695114-5G
Phenanthrene, Sigma-Aldrich®
Phenanthrene
Formula:
C₁₄H₁₀ Boiling Pt: 340 °C (1013 hPa) Melting Pt: 97…99 °C Density: 1.19 g/cm³ (20 °C) Flash Pt: 153 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001168 CAS Number: 85-01-8 EINECS: 201-581-5 UN: 3077 ADR: 9,III Merck Index: 12,07354 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...