Dimidium bromide, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 2,7-Diamino-10-methyl-9-phenylphenanthridinium bromide, 3,8-Diamino-5-methyl-6-phenylphenanthridinium bromide
41785-250MG 41785-1G
SIAL41785-250MGEA 163 EUR
SIAL41785-250MG SIAL41785-1G
Dimidium bromide, Sigma-Aldrich®
Dimidium bromide
Formula: C₂₀H₁₈BrN₃
Melting Pt: 243…248 °C
Storage Temperature: Ambient
MDL Number: MFCD00011757
CAS Number: 518-67-2
EINECS: 208-256-7
UN: 2811
ADR: 6.1,I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e669239c79726ce91f44ef372cbfed426e90f93b&vid=74b03720866852364cb9050de861a4f7c0e5e57b&prid=web&clid=SAPDAM