2-Methylpyrazine, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
M75608-100G
M75608-25G
SIALM75608-100GEA
49.4
EUR
SIALM75608-100G
SIALM75608-25G
2-Methylpyrazine, Sigma-Aldrich®
2-Methylpyrazine
Formula:
C₅H₆N₂ Boiling Pt: 135 °C (1013 hPa) Melting Pt: –30 °C Density: 1.03 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00006142 CAS Number: 109-08-0 EINECS: 203-645-8 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...