2,5-Dimethylpyrazine, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ300
W327204-100G-K W327204-1KG-K W327204-500G-K
SIALW327204-100G-KEA 95.9 EUR
SIALW327204-100G-K SIALW327204-1KG-K SIALW327204-500G-K
2,5-Dimethylpyrazine, Sigma-Aldrich®
2,5-Dimethylpyrazine
Formula: C₆H₈N₂
MW: 108.14 g/mol
Boiling Pt: 155
Melting Pt: 15
Density: 0.99 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006147
CAS Number: 123-32-0
EINECS: 204-618-3