2,6-Dimethylpyrazine, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ100
W327301-250G-K W327301-50G-K W327301-1KG-K
SIALW327301-250G-KEA 647 EUR
SIALW327301-250G-K SIALW327301-50G-K SIALW327301-1KG-K
2,6-Dimethylpyrazine, Sigma-Aldrich®
2,6-Dimethylpyrazine
Formula: C₆H₈N₂
MW: 108.14 g/mol
Boiling Pt: 154
Melting Pt: 38…42 °C
Density: 0.9647 g/cm³ (50 °C)
Flash Pt: 52
Storage Temperature: Ambient
MDL Number: MFCD00006148
CAS Number: 108-50-9
EINECS: 203-589-4
UN: 1325
ADR: 4.1 III